COMH4211AC05 Combi-Blocks, Inc.

5-Chloro-3-(4-methoxy-phenyl)-3h-[1,2,3]triazolo[4,5-d]pyrimidine

  • MDL number MFCD30830164
  • Cas Number : 1448694-38-9
  • Molecular Weight : 261.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1502
Availability details for
Quantity
  • Availability
  • Add to Cart