COMH4211AC06 Combi-Blocks, Inc.

N-(3,4-Dichloro-benzyl)-o-ethyl-hydroxylamine

  • MDL number MFCD21100552
  • Cas Number : 1457472-77-3
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1504

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1504
Availability details for
Quantity
  • Availability
  • Add to Cart