COMH4211AC07 Combi-Blocks, Inc.

[5-(2,5-Dichloro-phenyl)-thiophen-2-yl]-methanol

  • MDL number MFCD31619006
  • Cas Number : 2008825-96-3
  • Molecular Weight : 259.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1505

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1505
Availability details for
Quantity
  • Availability
  • Add to Cart