COMH4211AC0A Combi-Blocks, Inc.

1-(2-Methoxyethyl)piperidine-4-carbonitrile

  • MDL number MFCD11934584
  • Cas Number : 173974-82-8
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1515
Availability details for
Quantity
  • Availability
  • Add to Cart