COMH4211AC0B Combi-Blocks, Inc.

5-Bromo-2-chloro-n-(2,2-dimethoxyethyl)benzamide

  • MDL number MFCD13224162
  • Cas Number : 477772-67-1
  • Molecular Weight : 322.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1517
Availability details for
Quantity
  • Availability
  • Add to Cart