COMH4211AC0F Combi-Blocks, Inc.

3-Fluoro-4-(1,2,3,6-tetrahydro-pyridin-4-yl)-phenol hydrochloride

  • MDL number MFCD31583382
  • Cas Number : 2270912-31-5
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1524
Availability details for
Quantity
  • Availability
  • Add to Cart