COMH4211AC10 Combi-Blocks, Inc.

[(2-Bromo-3-methoxyphenyl)methyl](methyl)amine

  • MDL number MFCD26648005
  • Cas Number : 1509239-12-6
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1526

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1526
Availability details for
Quantity
  • Availability
  • Add to Cart