COMH4211AC11 Combi-Blocks, Inc.

1-[4-(Oxetan-3-yloxy)-phenyl]-ethanone

  • MDL number MFCD21848302
  • Cas Number : 1601779-56-9
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1528
Availability details for
Quantity
  • Availability
  • Add to Cart