COMH4211AC13 Combi-Blocks, Inc.

1-(2-Chloro-4-cyclopropylmethoxy-phenyl)-ethylamine

  • MDL number MFCD20296967
  • Cas Number : 1339184-46-1
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1531
Availability details for
Quantity
  • Availability
  • Add to Cart