COMH4211AC14 Combi-Blocks, Inc.

[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-carbamic acid 4-nitro-phenyl ester

  • MDL number MFCD31583145
  • Cas Number : 957198-19-5
  • Molecular Weight : 384.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1532
Availability details for
Quantity
  • Availability
  • Add to Cart