COMH4211AC16 Combi-Blocks, Inc.

(2R)-2-[(2,4-Dimethoxybenzyl)amino]propan-1-ol

  • MDL number MFCD31613915
  • Cas Number : 1604272-60-7
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1536

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1536
Availability details for
Quantity
  • Availability
  • Add to Cart