COMH4211AC19 Combi-Blocks, Inc.

4-Cyclopropylmethoxy-3-fluoro-benzylamine hydrochloride

  • MDL number MFCD31381716
  • Cas Number : 2206610-59-3
  • Molecular Weight : 231.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1541
Availability details for
Quantity
  • Availability
  • Add to Cart