COMH4211AC1A Combi-Blocks, Inc.

1-Piperidinecarboxylic acid, 4-(3-fluoro-4-nitrophenoxy)-, 1,1-dimethylethyl ester

  • MDL number MFCD30294759
  • Cas Number : 1232234-74-0
  • Molecular Weight : 340.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1543
Availability details for
Quantity
  • Availability
  • Add to Cart