COMH4211AC1B Combi-Blocks, Inc.

2-Chloro-4-isopropoxy-benzylamine

  • MDL number MFCD31539570
  • Cas Number : 1072065-14-5
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1544
Availability details for
Quantity
  • Availability
  • Add to Cart