COMH4211AC1C Combi-Blocks, Inc.

[3-(5-Amino-3-tert-butyl-pyrazol-1-yl)-phenyl]-methanol

  • MDL number MFCD28404256
  • Cas Number : 725686-00-0
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1545
Availability details for
Quantity
  • Availability
  • Add to Cart