COMH4211AC1F Combi-Blocks, Inc.

N-(1-Phenylethyl)piperidine-4-carboxamide

  • MDL number MFCD09809749
  • Cas Number : 73415-62-0
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1549

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1549
Availability details for
Quantity
  • Availability
  • Add to Cart