COMH4211AC20 Combi-Blocks, Inc.

4-(4-(2-tert-Butylphenyl)piperazin-1-yl)-4-oxobutanoic acid

  • MDL number MFCD31613744
  • Cas Number : 1252656-11-3
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1551
Availability details for
Quantity
  • Availability
  • Add to Cart