COMH4211AC21 Combi-Blocks, Inc.

C-(4-Chloro-2-fluoro-phenyl)-c-phenyl-methylamine hydrochloride

  • MDL number MFCD31557814
  • Cas Number : 2203140-45-6
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1552
Availability details for
Quantity
  • Availability
  • Add to Cart