COMH4211AC22 Combi-Blocks, Inc.

tert-Butyl 2,4-dichlorobenzoate

  • MDL number MFCD06291311
  • Cas Number : 220283-13-6
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1553
Availability details for
Quantity
  • Availability
  • Add to Cart