COMH4211AC23 Combi-Blocks, Inc.

5-(4-Cyclopropylmethoxy-2-fluoro-phenyl)-1h-tetrazole

  • MDL number MFCD31579405
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1558
Availability details for
Quantity
  • Availability
  • Add to Cart