COMH4211AC25 Combi-Blocks, Inc.

Methyl 5-hydroxy-2-methoxybenzoate

  • MDL number MFCD01940408
  • Cas Number : 87513-63-1
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1560
Availability details for
Quantity
  • Availability
  • Add to Cart