COMH4211AC2B Combi-Blocks, Inc.

O-(Naphthalen-1-ylmethyl)hydroxylamine

  • MDL number MFCD18971146
  • Cas Number : 54484-68-3
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1573

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1573
Availability details for
Quantity
  • Availability
  • Add to Cart