COMH4211AC2D Combi-Blocks, Inc.

6-(5-Bromo-2-methyl-benzyl)-2,3-dihydro-benzo[1,4]dioxine

  • MDL number MFCD31579379
  • Cas Number : 1673515-05-3
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1588
Availability details for
Quantity
  • Availability
  • Add to Cart