COMH4211AC30 Combi-Blocks, Inc.

5-Bromo-2-methoxy-n-methylbenzamide

  • MDL number MFCD08104771
  • Cas Number : 899143-67-0
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1601
Availability details for
Quantity
  • Availability
  • Add to Cart