COMH4211AC32 Combi-Blocks, Inc.

5-Chloro-2-(morpholin-4-yl)benzaldehyde

  • MDL number MFCD28967680
  • Cas Number : 1446818-96-7
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1605
Availability details for
Quantity
  • Availability
  • Add to Cart