COMH4211AC35 Combi-Blocks, Inc.

(5-Methyl-3-(m-tolyl)isoxazol-4-yl)methanol

  • MDL number MFCD19349006
  • Cas Number : 1159602-00-2
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1608

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1608
Availability details for
Quantity
  • Availability
  • Add to Cart