COMH4211AC36 Combi-Blocks, Inc.

Di(n-succinimidyl) adipate

  • MDL number MFCD01318516
  • Cas Number : 59156-70-6
  • Molecular Weight : 340.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-({6-[(2,5-dioxo-1-pyrrolidinyl)oxy]-6-oxohexanoyl}oxy)-2,5-pyrrolidinedione
Inchi Code 1S/C14H16N2O8/c17-9-5-6-10(18)15(9)23-13(21)3-1-2-4-14(22)24-16-11(19)7-8-12(16)20/h1-8H2
Inchi Key LZZXZDMVRZJZST-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1609
Availability details for
Quantity
  • Availability
  • Add to Cart