COMH4211AC37 Combi-Blocks, Inc.

4-Bromo-6-chloro-2,3-difluoroaniline

  • MDL number MFCD31583208
  • Cas Number : 1895912-80-7
  • Molecular Weight : 242.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1610

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1610
Availability details for
Quantity
  • Availability
  • Add to Cart