COMH4211AC39 Combi-Blocks, Inc.

1-(Cyclopropylmethyl)-1,2,3,4-tetrahydroquinolin-7-amine

  • MDL number MFCD16702210
  • Cas Number : 1250681-39-0
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1614
Availability details for
Quantity
  • Availability
  • Add to Cart