COMH4211AC3B Combi-Blocks, Inc.

3-Bromo-4-(2,2,2-trifluoroethoxy)benzonitrile

  • MDL number MFCD25801480
  • Cas Number : 1065640-69-8
  • Molecular Weight : 280.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1616

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1616
Availability details for
Quantity
  • Availability
  • Add to Cart