COMH4211AC3D Combi-Blocks, Inc.

2-(3-Iodophenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD12463391
  • Cas Number : 1097101-30-8
  • Molecular Weight : 379.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-iodophenoxy)-5-(trifluoromethyl)aniline
Inchi Code 1S/C13H9F3INO/c14-13(15,16)8-4-5-12(11(18)6-8)19-10-3-1-2-9(17)7-10/h1-7H,18H2
Inchi Key IWNHRUSVEJGMBA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1623
Availability details for
Quantity
  • Availability
  • Add to Cart