COMH4211AC40 Combi-Blocks, Inc.

Acetamide, n-[1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]-2-methylpropyl]-, (s)-

  • MDL number MFCD27965336
  • Cas Number : 56186-37-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1634

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1634
Availability details for
Quantity
  • Availability
  • Add to Cart