COMH4211AC44 Combi-Blocks, Inc.

2-(3-Benzyltriazol-4-yl)ethanol

  • MDL number MFCD31579385
  • Cas Number : 1619971-58-2
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1641
Availability details for
Quantity
  • Availability
  • Add to Cart