COMH4211AC45 Combi-Blocks, Inc.

(R)-1-(1-Phenylethyl)urea

  • Cas Number : 16849-91-5
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[(1R)-1-phenylethyl]urea
Inchi Code 1S/C9H12N2O/c1-7(11-9(10)12)8-5-3-2-4-6-8/h2-7H,1H3,(H3,10,11,12)/t7-/m1/s1
Inchi Key ALVBVEKNACPPAB-SSDOTTSWSA-N

Description

Purity 95%
Country of Origin CN
Availability details for
Quantity
  • Availability
  • Add to Cart