COMH4211AC47 Combi-Blocks, Inc.

3-(4-Cyclopropylmethoxy-phenyl)-propionic acid

  • MDL number MFCD17171054
  • Cas Number : 1047680-56-7
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1647
Availability details for
Quantity
  • Availability
  • Add to Cart