COMH4211AC48 Combi-Blocks, Inc.

(4-Methoxymethyl-bicyclo[2.2.2]oct-1-yl)-methanol

  • MDL number MFCD31381611
  • Cas Number : 2167066-70-6
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1650
Availability details for
Quantity
  • Availability
  • Add to Cart