COMH4211AC4B Combi-Blocks, Inc.

2-[4-(4-Chlorophenyl)piperazin-1-yl]acetic acid

  • MDL number MFCD11164475
  • Cas Number : 946757-68-2
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1654
Availability details for
Quantity
  • Availability
  • Add to Cart