COMH4211AC4C Combi-Blocks, Inc.

1-(Piperidin-1-ylmethyl)cyclopropane-1-carboxylic acid hydrochloride

  • MDL number MFCD31381709
  • Cas Number : 2204587-97-1
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1656
Availability details for
Quantity
  • Availability
  • Add to Cart