COMH4211AC51 Combi-Blocks, Inc.

4-(Trifluoromethoxy)biphenyl-2-amine

  • MDL number MFCD31557805
  • Cas Number : 1628909-04-5
  • Molecular Weight : 253.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1664
Availability details for
Quantity
  • Availability
  • Add to Cart