COMH4211AC52 Combi-Blocks, Inc.

1-(3-Bromo-5-fluorophenyl)ethylamine

  • MDL number MFCD18660092
  • Cas Number : 1191908-56-1
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1666

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1666
Availability details for
Quantity
  • Availability
  • Add to Cart