COMH4211AC55 Combi-Blocks, Inc.

5-Amino-2-ethoxybenzamide

  • MDL number MFCD11695276
  • Cas Number : 81929-48-8
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1670
Availability details for
Quantity
  • Availability
  • Add to Cart