COMH4211AC57 Combi-Blocks, Inc.

3-Bromo-4-(pyrrolidin-1-ylmethyl)pyridine

  • MDL number MFCD23163979
  • Cas Number : 1260009-55-9
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1674
Availability details for
Quantity
  • Availability
  • Add to Cart