COMH4211AC5A Combi-Blocks, Inc.

1-(4-Chloro-2-methoxyphenyl)ethan-1-ol

  • MDL number MFCD30718696
  • Cas Number : 1593370-55-8
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1677
Availability details for
Quantity
  • Availability
  • Add to Cart