COMH4211AC5C Combi-Blocks, Inc.

4-[(4H-1,2,4-Triazol-4-yl)amino]benzonitrile

  • MDL number MFCD12405686
  • Cas Number : 148869-66-3
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1681
Availability details for
Quantity
  • Availability
  • Add to Cart