COMH4211AC5D Combi-Blocks, Inc.

1-(2-Fluoro-4-methyl-benzyl)-piperazine dihydrochloride

  • MDL number MFCD31557906
  • Cas Number : 2208779-90-0
  • Molecular Weight : 281.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1682
Availability details for
Quantity
  • Availability
  • Add to Cart