COMH4211AC5E Combi-Blocks, Inc.

2-(2-(tert-Butyldiphenylsilyloxy)ethyl)aniline

  • MDL number MFCD28967464
  • Cas Number : 931105-21-4
  • Molecular Weight : 375.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1683
Availability details for
Quantity
  • Availability
  • Add to Cart