COMH4211AC60 Combi-Blocks, Inc.

(2,3,6-Trichlorophenyl)methanamine

  • MDL number MFCD07786738
  • Cas Number : 4960-49-0
  • Molecular Weight : 210.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1692

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1692
Availability details for
Quantity
  • Availability
  • Add to Cart