COMH4211AC61 Combi-Blocks, Inc.

2-(2-(Methylthio)ethoxy)propanoic acid

  • MDL number MFCD20645907
  • Cas Number : 870562-01-9
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1693
Availability details for
Quantity
  • Availability
  • Add to Cart