COMH4211AC62 Combi-Blocks, Inc.

4-Allyloxy-1-(tetrahydro-pyran-2-yl)-1h-pyrazole

  • MDL number MFCD31579306
  • Cas Number : 2270910-93-3
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1694
Availability details for
Quantity
  • Availability
  • Add to Cart