COMH4211AC66 Combi-Blocks, Inc.

2,5-Dibromo-3-ethoxypyridine

  • MDL number MFCD00234289
  • Cas Number : 887571-07-5
  • Molecular Weight : 280.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1700
Availability details for
Quantity
  • Availability
  • Add to Cart